Learn More
Epredia™ TissueLoc™ Cassette IV Biopsy Cassettes
Product Code. 16384156 Shop All Epredia Products
Change view
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 1000
Product Code. Color unitSize
16384156 Beige Pack of 1000
16394156 Blue Pack of 1000
16304166 Green Pack of 1000
16314166 Gray Pack of 1000
16324166 Lavender Pack of 1000
16334166 Light Green Pack of 1000
16344166 Lilac Pack of 1000
16354166 Orange Pack of 1000
16364166 Pink Pack of 1000
16374166 Tan Pack of 1000
16384166 White Pack of 1000
16394166 Yellow Pack of 1000
12 options
Product Code. 16384156
Brand: Epredia™ B851925BE
For biopsy tissue samples, these cassettes have a back-edge lid hinge with loose-packed lids that are available in 12 different color varieties
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Small Squares
Closure Included Yes
Quantity 1000/Cs.
Closure Type Separate Lid, Back Hinge
PrintMate Compatible No
Color Beige
Product Type TissueLoc Cassette IV with Color-Matched Lids