Learn More
Kimble Chase™ Replacement PTFE Pestle for Kimble™ Kontes™ DUALL™ Tissue Ginder, Size 20
Product Code. 10569792
Change view
Click to view available options
Capacity (Metric):
1 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
10569792 1 mL Each
1 options
Product Code. 10569792
Brand: Kimble Chase™ 8854810020
Only null left
Add to basket
Add to basket
The DUALL™ pestle works to combine both conical and cylindrical surfaces to effectively reduce tissue and produce a uniform homogenate.
  • Manufactured from borosilicate glass 3.3
  • Ideal for soft tissue such as brain or liver
  • Strong construction allows the pestle to be motor-driven
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Ideal for soft tissue such as brain and liver
For Use With (Equipment) Kimble DUALL Tissue Grinder
Capacity (Metric) 1 mL
Length (Metric) 155 mm
Material PTFE
Size 20
Product Type Pestle