Learn More
TPP™ Tissue Culture Flasks
Click to view available options
Volume (Metric) Working:
15 mL to 45 mL
15 to 45 mL
3 mL to 8 mL
30 to 85 mL
Unit Size:
Pack of 100
Pack of 18
Pack of 36
Pack of 360
Description
Surface activated, uniform flat growth surface with crystal clear transparency
- Surface activated, uniform flat growth surface
- Crystal clear transparency
- Geometry of neck reduces risk of media slop over
- Stable, slip-free stacking of multiple TPP flasks
- Highly visible bilateral marking area
- White and accurate volume graduations for optic control of the filling volume
Specifications
Specifications
| Description | Dimensions: 275 x 170 x 50 mm |
| Material | Polystyrene |
| No. per Case | 18 |
| Graduated | Yes |
| Bottom Shape | Flat |
| Culture Area | 300 cm2 |
| Volume (Metric) Working | 30 to 85 mL |