Learn More
Tirofiban hydrochloride monohydrate, MedChemExpress
Product Code. 16487308
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16487308
Brand: Medchem Express HY17369
MedChemExpress Tirofiban (L700462) hydrochloride monohydrate is a selective and reversible platelet integrin receptor (Gp IIb/IIIa) antagonist that inhibits fibrinogen binding to this receptor and has antithrombotic activity.
Tirofiban hydrochloride monohydrate induces proliferation and migration on endothelial cell by inducing production of VEGF. Tirofiban hydrochloride monohydrate can significantly reduces myocardial no-reflow and ischemia-reperfusion injury by alleviating myocardial microvascular structural and endothelial dysfunction in the ischemic area.
Specifications
Chemical Name or Material Tirofiban hydrochloride monohydrate
CAS 150915-40-5
Color White
Purity Grade Notes Research
Molecular Formula C22H39ClN2O6S
Quantity 10 mg
Synonym L700462 hydrochloride monohydrateMK383 hydrochloride monohydrate
Solubility Information DMSO : 100 mg/mL (201.99 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.02 mM; Need ultrasonic)
SMILES O=S(N[C@H](C(O)=O)CC1=CC=C(OCCCCC2CCNCC2)C=C1)(CCCC)=O.[H]Cl.[H]O[H]
Molecular Weight (g/mol) 495.07
Formula Weight 495.07
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.