missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tirofiban hydrochloride monohydrate, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
Tirofiban hydrochloride monohydrate induces proliferation and migration on endothelial cell by inducing production of VEGF. Tirofiban hydrochloride monohydrate can significantly reduces myocardial no-reflow and ischemia-reperfusion injury by alleviating myocardial microvascular structural and endothelial dysfunction in the ischemic area.
Specifications
Specifications
| Chemical Name or Material | Tirofiban hydrochloride monohydrate |
| CAS | 150915-40-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H39ClN2O6S |
| Quantity | 100 mg |
| Synonym | L700462 hydrochloride monohydrateMK383 hydrochloride monohydrate |
| Solubility Information | DMSO : 100 mg/mL (201.99 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.02 mM; Need ultrasonic) |
| SMILES | O=S(N[C@H](C(O)=O)CC1=CC=C(OCCCCC2CCNCC2)C=C1)(CCCC)=O.[H]Cl.[H]O[H] |
| Molecular Weight (g/mol) | 495.07 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction