Learn More
- Home
- Products
- Environmental Samplers
- Particulate Samplers
- Sampling Systems™ Tip for Unit Dose Sleeve Sampler
Sampling Systems™ Tip for Unit Dose Sleeve Sampler
Product Code. 15879281 Shop All Sampling Systems Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Length (Metric):
9.5 mm
11.5 mm
13.5 mm
14.5 mm
15.5 mm
17.5 mm
18.5 mm
19.5 mm
21.5 mm
22.5 mm
23.5 mm
25.5 mm
26.5 mm
27.5 mm
29.5 mm
30.5 mm
31.5 mm
34.5 mm
35.5 mm
38 mm
41.5 mm
45.5 mm
49 mm
52.5 mm
54 mm
Capacity (Metric):
0.1 mL
0.2 mL
0.3 mL
0.4 mL
0.5 mL
0.6 mL
0.7 mL
0.8 mL
0.9 mL
1.0 mL
1.1 mL
1.2 mL
1.3 mL
1.4 mL
1.5 mL
1.6 mL
1.7 mL
1.8 mL
1.9 mL
2 mL
2.1 mL
2.2 mL
2.3 mL
2.4 mL
2.5 mL
Diameter (Metric):
6 mm
8 mm
missing translation for 'unitSize'
Each
| Product Code. | Length (Metric) | Capacity (Metric) | Diameter (Metric) | unitSize |
|---|---|---|---|---|
| 15879281 | 17.5 mm | 2.1 mL | 8 mm | Each |
| 15809271 | 45.5 mm | 0.4 mL | 6 mm | Each |
| 15809281 | 31.5 mm | 1.4 mL | 8 mm | Each |
| 15809291 | 11.5 mm | 2.4 mL | 8 mm | Each |
| 15819271 | 41.5 mm | 0.5 mL | 6 mm | Each |
| 15819281 | 29.5 mm | 1.5 mL | 8 mm | Each |
| 15819291 | 9.5 mm | 2.5 mL | 8 mm | Each |
| 15829271 | 38 mm | 0.6 mL | 6 mm | Each |
| 15829281 | 27.5 mm | 1.6 mL | 8 mm | Each |
| 15839271 | 34.5 mm | 0.7 mL | 6 mm | Each |
| 15839281 | 25.5 mm | 1.7 mL | 8 mm | Each |
| 15849271 | 30.5 mm | 0.8 mL | 6 mm | Each |
| 15849281 | 23.5 mm | 1.8 mL | 8 mm | Each |
| 15859271 | 26.5 mm | 0.9 mL | 6 mm | Each |
| 15859281 | 21.5 mm | 1.9 mL | 8 mm | Each |
| 15869271 | 22.5 mm | 1.0 mL | 6 mm | Each |
| 15869281 | 19.5 mm | 2 mL | 8 mm | Each |
| 15879261 | 54 mm | 0.1 mL | 6 mm | Each |
| 15879271 | 18.5 mm | 1.1 mL | 6 mm | Each |
| 15889261 | 52.5 mm | 0.2 mL | 6 mm | Each |
| 15889271 | 14.5 mm | 1.2 mL | 6 mm | Each |
| 15889281 | 15.5 mm | 2.2 mL | 8 mm | Each |
| 15899261 | 49 mm | 0.3 mL | 6 mm | Each |
| 15899271 | 35.5 mm | 1.3 mL | 8 mm | Each |
| 15899281 | 13.5 mm | 2.3 mL | 8 mm | Each |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- For sampling small volume powders and granules.
- Chemically resistant 316L stainless steel construction.
- Different sample sizes can be taken using the same sampler
- Tips for use with Type 1 or Type 2 Sampling Chambers
Specifications
Specifications
| Product Type | Unit Dose Sleeve Sampler Tip |
| Material | Stainless Steel (316L) |
| Capacity (Metric) | 2.1 mL |
| Diameter (Metric) | 8 mm |
| Length (Metric) | 17.5 mm |