Learn More
Sampling Systems™ Tip for Unit Dose Sleeve Sampler
Product Code. 15849281 Shop All Sampling Systems Products
Change view
Click to view available options
Length (Metric):
9.5 mm
11.5 mm
13.5 mm
14.5 mm
15.5 mm
17.5 mm
18.5 mm
19.5 mm
21.5 mm
22.5 mm
23.5 mm
25.5 mm
26.5 mm
27.5 mm
29.5 mm
30.5 mm
31.5 mm
34.5 mm
35.5 mm
38 mm
41.5 mm
45.5 mm
49 mm
52.5 mm
54 mm
Capacity (Metric):
0.1 mL
0.2 mL
0.3 mL
0.4 mL
0.5 mL
0.6 mL
0.7 mL
0.8 mL
0.9 mL
1.0 mL
1.1 mL
1.2 mL
1.3 mL
1.4 mL
1.5 mL
1.6 mL
1.7 mL
1.8 mL
1.9 mL
2 mL
2.1 mL
2.2 mL
2.3 mL
2.4 mL
2.5 mL
Diameter (Metric):
6 mm
8 mm
Unit Size:
Each
Product Code. Length (Metric) Capacity (Metric) Diameter (Metric) unitSize
15849281 23.5 mm 1.8 mL 8 mm Each
15809271 45.5 mm 0.4 mL 6 mm Each
15809281 31.5 mm 1.4 mL 8 mm Each
15809291 11.5 mm 2.4 mL 8 mm Each
15819271 41.5 mm 0.5 mL 6 mm Each
15819281 29.5 mm 1.5 mL 8 mm Each
15819291 9.5 mm 2.5 mL 8 mm Each
15829271 38 mm 0.6 mL 6 mm Each
15829281 27.5 mm 1.6 mL 8 mm Each
15839271 34.5 mm 0.7 mL 6 mm Each
15839281 25.5 mm 1.7 mL 8 mm Each
15849271 30.5 mm 0.8 mL 6 mm Each
15859271 26.5 mm 0.9 mL 6 mm Each
15859281 21.5 mm 1.9 mL 8 mm Each
15869271 22.5 mm 1.0 mL 6 mm Each
15869281 19.5 mm 2 mL 8 mm Each
15879261 54 mm 0.1 mL 6 mm Each
15879271 18.5 mm 1.1 mL 6 mm Each
15879281 17.5 mm 2.1 mL 8 mm Each
15889261 52.5 mm 0.2 mL 6 mm Each
15889271 14.5 mm 1.2 mL 6 mm Each
15889281 15.5 mm 2.2 mL 8 mm Each
15899261 49 mm 0.3 mL 6 mm Each
15899271 35.5 mm 1.3 mL 8 mm Each
15899281 13.5 mm 2.3 mL 8 mm Each
25 options
Product Code. 15849281
Brand: Sampling Systems™ 1500W18
Only null left
Add to basket
Add to basket
Developed for customers requiring accurate unit dose samples in small volumes. Sampling Systems™ Tip for Unit Dose Sleeve Sampler is ideal for use with a wide range of free flowing powders and granules.
  • For sampling small volume powders and granules.
  • Chemically resistant 316L stainless steel construction.
  • Different sample sizes can be taken using the same sampler
  • Tips for use with Type 1 or Type 2 Sampling Chambers
Specifications
Product Type Unit Dose Sleeve Sampler Tip
Material Stainless Steel (316L)
Capacity (Metric) 1.8 mL
Diameter (Metric) 8 mm
Length (Metric) 23.5 mm