missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Tintometer™ VARIO™ Hydrazine Reagent Solution, 100mL
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
- Range: 0.01 to 0.6mg/L
- Parameter: Hydrazine
- Method: Dimethylamino-benzaldehyde
- For use in test kits, colorimeters and spectrophotometers
- Sold individually
Specifications
Specifications
| For Use With (Application) | Water Testing |
| For Use With (Equipment) | Test kits, colorimeters and spectrophotometers |
| Quantity | 100 mL |
| Test Method | Dimethylamino-benzaldehyde |
| Type | VARIO™ Hydrazine Reagent Solution |
Safety and Handling
Product Identifier
- SOLUTION VARIO HYDRA 2 REAGENT
- Danger
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P390-Absorb spillage to prevent material damage.
- MIXTURE LIST-Contains : sulphuric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction