Learn More
Lovibond™ Tintometer™ SensoDirect 150 All-In-One Handheld Meter, pH/Con/TDS/Oxi
Description
Features include automatic calibration, buffer recognition, large LCD display and result hold function. Lovibond SensoDirect 150 is combined instrument capable of measuring pH, ORP, Conductivity, TDS, Dissolved Oxygen and Temperature in a single meter.
- Test: pH Value; Range: 0 to 14
- Test: Conductivity; Range: 0 to 200uS/cm; 0.2 to 200mS/cm
- Test: Dissolved Oxygen; Range: 0 to 20mg/L
- Includes:
- SensoDirect 150 meter
- Battery, 9V
- pH electrode, plastic, type 110
- Conductivity Electrode
- Dissolved Oxygen Sensor
- pH buffer set with pH 4.0 and pH 7
- Electrolyte solution for DO sensor
- Replacement DO sensor heads
- Instruction manual and guarentee sheet
Specifications
Specifications
| Accuracy (pH) | 0 to 14 pH, ±0.02 pH + 2 digits |
| Accuracy (mV) | 0 to 1999mV, 1mV |
| Accuracy (Temperature) | ±0.8°C (1.5°F) |
| Type | SensoDirect 150 Set pH/Con/TDS/Oxi |
| Dimensions (L x W x H) | 220 x 120 x 40 mm |
| For Use With (Application) | Measures pH/Redox, dissolved oxygen and conductivity/TDS |
| pH Range | 0 to 14 |
| Range (Conductivity) | 0 to 200.0 µS |
| Range (mV) | mV to 1999mV to 1999mV |
| Range (TDS) | 0 to 132,000ppm |
| Show More |