Learn More
Lovibond™ Tintometer™ Refill Reagent Packs, Biguanide / pH Tests
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Brand:  Lovibond™ 516155
Additional Details : Weight : 0.30000kg
Product Code. 15361905
  • 238.00 NOK / Each
Estimated Shipment: 02-07-2024
to see stock.
Add to basket
Description
Description
  • Contains:
  • 30 PHMB Tablets
  • 30 Phenol Red Rapid Tablets