Learn More
Lovibond™ Tintometer™ Refill Reagent Packs, Biguanide / pH Tests
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Brand: Lovibond™ 516155
Additional Details : Weight : 0.30000kg
Description
- Contains:
- 30 PHMB Tablets
- 30 Phenol Red Rapid Tablets