Learn More
Lovibond™ Tintometer™ Molybdate No. 1 HR / No. 2 HR Tablets Combi Pack
Product Code. 15341915
Change view
Click to view available options
Quantity:
250 Tablets
500 Tablets
Unit Size:
Pack of 100
Pack of 250
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15341915 500 Tablets Pack of 250
15331915 250 Tablets Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15341915 Supplier Lovibond™ Supplier No. 517632BT
  • ONLINE EXCLUSIVE PRICE
    2370.00 NOK
    1118.77 NOK / Pack of 250
    Save 1251.23 NOK
    52% Off
 Restricted Item
Estimated Shipment: 30-11-2026
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
  • Range: 1 to 50mg/L
  • Parameter: Molybdate
  • Method: Thioglycolate
  • New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
  • With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
  • Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
  • Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
  • ‘BT’ is added to the end of the code to identify the new style of packaging
  • Simple, time-saving and practical
  • There are no safety risks if the tablets are used in line with the instructions supplied
  • Safety data sheets are available for all reagents
Specifications
For Use With (Application) Photometry Tablet Reagents
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Quantity 500 Tablets
Range 1 to 50mg/L
Type Molybdate No. 1 HR / No. 2 HR Tablets Combi Pack: 2500 ea
compliance-icons compliance-icons compliance-icons
Product Identifier
  • X250 COMBI-PACK MOLYBDATE HR NO. 1 + HR NO. 2 .
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Adipic acid, Sodiumbisulphate, Potassium persulphate, Calcium thioglycolate trihydrate