Learn More
Lovibond™ Tintometer™ Methyl Red Solution, 100mL
Product Code. 15342025
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15342025 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15342025 Supplier Lovibond™ Supplier No. 451631
  • ONLINE EXCLUSIVE PRICE
    407.00 NOK
    360.24 NOK / 100mL
    Save 46.76 NOK
    11% Off
 Restricted Item
Estimated Shipment: 14-12-2026
For use with Lovibond test kits, colorimeters and spectrophotometers
  • Methyl Red Solution
  • 100mL
Specifications
For Use With (Equipment) For use with Lovibond test kits, colorimeters and spectrophotometers
Quantity 100 mL
Type Methyl Red Solution
compliance-icons
Product Identifier
  • 100ML METHYLRED
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contain: propan-2-ol