Learn More
Tintometer™ Lovibond™ Water Testing Reagents
Product Code. 13223459
Change view
Click to view available options
Quantity:
100/Pk.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13223459 100/Pk. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13223459 Supplier Tintometer™ Supplier No. 530550
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use in water testing
  • Iron Vario TPTZ F10
  • Available as powder pack
  • For water, waste water, seawater testing
Specifications
For Use With (Application) For water testing, waste water testing, seawater testing
Quantity 100/Pk.
Size 100/Pk.
Type Iron vario TPTZ F10 reagent powder
compliance-icons
Product Identifier
  • Vario TPTZ Iron F10Ml
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • EUH031-Contact with acids liberates toxic gas.
  • EUH208-Contains methenamine. May produce an allergic reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: disodium disulphite,sodium dithionite,methenamine