Learn More
Lovibond™ Tintometer™ DPD No. 4 Rapid Tablet
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Brand:  Lovibond™ 511572BT
Additional Details : Weight : 0.30000kg
 View more versions of this product
Product Code. 15381955
  • 1070.00 NOK / Pack of 500
Estimated Shipment: 22-07-2024
Description
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
  • DPD No. 4 Rapid Tablet
  • New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
  • With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
  • Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
  • Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
  • ‘BT’ is added to the end of the code to identify the new style of packaging
  • Simple, time-saving and practical
  • There are no safety risks if the tablets are used in line with the instructions supplied
  • Safety data sheets are available for all reagents
Specifications
Specifications
Photometry Tablet Reagents
Test kits, colorimeters and spectrophotometers
500 Tablets
DPD No. 4 Rapid Tablet