Learn More
Lovibond™ Tintometer™ Boron No. 1 Tablet, 0.1 to 2mg/L
Click to view available options
Quantity:
100 Tablets
Unit Size:
Pack of 100
Description
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
- Range: 0.1 to 2mg/L
- Parameter: Boron
- Method: Azomethine
- For use in test kits, colorimeters and spectrophotometers
- Case of 100
Specifications
Specifications
| For Use With (Application) | Photometry Tablet Reagents |
| For Use With (Equipment) | Test kits, colorimeters and spectrophotometers |
| Quantity | 100 Tablets |
| Range | 0.1 to 2mg/L |
| Test Method | Azomethine |
| Type | Boron No. 1 Tablets |