Learn More
Lovibond™ Tintometer™ Boron No. 1 Tablet, 0.1 to 2mg/L
Product Code. 13455089
Click to view available options
Quantity:
100 Tablets
Unit Size:
Pack of 100
Product Code. 13455089
Brand: Lovibond™ 515790BT
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
  • Range: 0.1 to 2mg/L
  • Parameter: Boron
  • Method: Azomethine
  • For use in test kits, colorimeters and spectrophotometers
  • Case of 100
Specifications
For Use With (Application) Photometry Tablet Reagents
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Quantity 100 Tablets
Range 0.1 to 2mg/L
Test Method Azomethine
Type Boron No. 1 Tablets