missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Tintometer™ Aqua Dest Distilled Water, 250mL

Product Code. 15332035
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
This item is not returnable. View return policy

Product Code. 15332035

Brand: Lovibond™ 457022

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use with Lovibond test kits, colorimeters and spectrophotometers

  • Aqua Dest
  • Distilled Water
  • 250mL

Specifications

For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Quantity 250 mL
Type Aqua Dest Distilled Water
Product Identifier
  • 250ML AQUA DEST.
Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid, N,N-diethylbenzene-1,4-diammonium sulphate,butan-2-ol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.