missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TINAGL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | TINAGL1 |
---|---|
Concentration | 0.17 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16899573
|
Proteintech
12077-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16889573
|
Proteintech
12077-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
TINAGL1 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
Q99JR5, Q9EQT5, Q9GZM7 | |
64129, 94174, 94242 | |
TINAGL1 Fusion Protein Ag2721 | |
Primary | |
-20°C | |
TINAGL1 |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ARG1, GIS5, LCN7, LIECG3, OLRG 2, OLRG2, PSEC0088, TIN Ag related protein, TIN Ag RP, TINAGL, TINAGL1, TINAGRP, UNQ204/PRO230 | |
TINAGL1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |