Learn More
Tin(IV) Oxide, Extra Pure, MilliporeSigma™
Product Code. 15875050
Change view
Click to view available options
Quantity:
250 g
25 kg
Packaging:
Fibre carton
Plastic bottle
Unit Size:
250g
25kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15875050 250 g Plastic bottle 250g
15805060 25 kg Fibre carton 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15875050 Supplier Supelco Supplier No. 1078180250
 Restricted Item
Estimated Shipment: 26-11-2026
Chemical Identifiers
CAS 18282-10-5
Molecular Formula O2Sn
Molecular Weight (g/mol) 150.71
MDL Number MFCD00011244
InChI Key XOLBLPGZBRYERU-UHFFFAOYSA-N
Synonym tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride
PubChem CID 29011
ChEBI CHEBI:52991
IUPAC Name stannanedione
SMILES O=[Sn]=O
Specifications
Physical Form Solid
Quantity 250 g
Odor Odorless
Packaging Plastic bottle
Appearance Off-white solid
Formula Weight 150.69
Percent Purity ≥99.0%
Grade Extra Pure
Chemical Name or Material Tin(IV) oxide