Learn More
Tin(II) chloride dihydrate, Supelco™
Product Code. 32237592
Change view
Click to view available options
Quantity:
25 kg
Unit Size:
25kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32237592 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32237592 Supplier Supelco Supplier No. 1078159025
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
For analysis EMSURE™ ACS,ISO,Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Specifications
Boiling Point 623°C (1013 hPa) (anhydrous substance)
Chemical Name or Material Tin(II) chloride dihydrate
Melting Point 38°C (anhydrous substance)
CAS 10025-69-1
Quantity 25 kg
Density 2.71 g/cm³ at 20°C
Synonym Tin(II) chloride dihydrate, Tin dichloride, dihydrate, Stannous chloride, Tin(II) chloride dihydrate
Molecular Weight (g/mol) 225.65
Grade Not Available