Learn More
Fisherbrand™ Quick-Timer™
Shop All Fisherbrand Products
Click to view available options
Time Range:
0 to 60 min.
Unit Size:
Each
Description
- Adjustable from 0 to 60 minutes in 1-minute increments
- Easy-to-read and use
- No keys, batteries, or confusing settings
- Case is chemical resistant ABS plastic
Specifications
Specifications
| Product Type | Timer |
| Time Range | 0 to 60 min. |
| Color | Blue/White |
| Weight (Metric) | 85.04 g |