Specifications
| CAS | 137330-13-3 |
| Melting Point | 130.0°C to 135.0°C |
| Molecular Formula | C46H83N2O17P |
| Quantity | 1 g |
| InChI Key | NESIVXZOSKKUDP-ARVJLQODSA-N |
| SMILES | OP(O)(O)=O.CC[C@H]1OC(=O)C[C@@H](O)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)[C@@H](O)[C@@H]([C@H]2O)N(C)C)[C@@H](CCN2C[C@@H](C)C[C@@H](C)C2)C[C@@H](C)C(=O)\C=C\C(\C)=C\[C@@H]1CO[C@@H]1O[C@H](C)[C@@H](O)[C@@H](OC)[C@H]1OC |
| IUPAC Name | (4R,5S,6S,7R,9R,11E,13E,15R,16R)-6-{[(2R,3R,4S,5S,6R)-4-(dimethylamino)-3,5-dihydroxy-6-methyloxan-2-yl]oxy}-7-{2-[(3R,5S)-3,5-dimethylpiperidin-1-yl]ethyl}-16-ethyl-4-hydroxy-15-({[(2R,3R,4R,5R,6R)-5-hydroxy-3,4-dimethoxy-6-methyloxan-2-yl]oxy}methyl)-5,9,13-trimethyl-1-oxacyclohexadeca-11,13-diene-2,10-dione; phosphoric acid |
| Molecular Weight (g/mol) | 967.14 |
| Formula Weight | 967.13 |
| Chemical Name or Material | Tilmicosin phosphate |