Learn More
Tilmicosin phosphate, MedChemExpress
Product Code. 16488496
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. 16488496
Brand: Medchem Express HYB0905A
MedChemExpress Tilmicosin phosphate is a antibiotic, is used for the research of bovine respiratory disease and ovine respiratory disease associated with Mannheimia (Pasteurella) haemolytica.
Tilmicosin phosphate is a antibiotic, is used for the research of bovine respiratory disease and ovine respiratory disease associated with Mannheimia (Pasteurella) haemolytica.
Specifications
Chemical Name or Material Tilmicosin phosphate
Physical Form Solid
CAS 137330-13-3
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C46H83N2O17P
For Use With (Application) COVID-19-immunoregulation
Synonym LY-177370 phosphate EL-870 phosphate
Solubility Information DMSO : ≥ 100 mg/mL (103.40 mM) ∣H2O : ≥ 100 mg/mL (103.40 mM)
SMILES C[C@@H]([C@@H](CC1=O)O)[C@H]([C@H](C[C@H](C(/C=C/C(C)=C/[C@H](CO[C@H](O[C@H](C)[C@@H](O)[C@H]2OC)[C@@H]2OC)[C@@H](CC)O1)=O)C)CCN(C[C@H](C)C3)C[C@H]3C)O[C@@](O[C@H](C)[C@@H](O)[C@@H]4N(C)C)([H])[C@@H]4O.O=P(O)(O)O
Molecular Weight (g/mol) 967.13
Formula Weight 967.13
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.