Learn More
BOLA Tiles
Product Code. 10004840
Change view
Click to view available options
Dimensions (L x W):
300 x 2 mm
300 x 3 mm
300 x 4 mm
300 x 5 mm
300 x 6 mm
300 x 8 mm
300 x 10 mm
300 x 15 mm
Unit Size:
Each
Product Code. Dimensions (L x W) unitSize
10004840 300 x 5 mm Each
10350671 300 x 2 mm Each
10116550 300 x 3 mm Each
10083531 300 x 4 mm Each
10401001 300 x 6 mm Each
10640562 300 x 8 mm Each
10605991 300 x 10 mm Each
10691132 300 x 15 mm Each
8 options
Product Code. 10004840
Brand: BOLA S180508
Only null left
Add to basket
Add to basket
Tiles virgin PTFE
Standard sizes with different thicknesses.
Specifications
Absorbancy Non-absorbent
Dimensions (L x W) 300 x 5 mm