missing translation for 'onlineSavingsMsg'
Learn More

TIGAR/C12orf5 Antibody [Janelia Fluor™ 635], Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP1-49534JF635

Product Code. 30170894

  • 5090.00 NOK / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TIGAR/C12orf5 Polyclonal antibody specifically detects TIGAR/C12orf5 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TIGAR/C12orf5
Polyclonal
50mM Sodium Borate
Rabbit
Immunogen affinity purified
RUO
Primary
Human, Mouse
Purified
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Janelia Fluor 635
chromosome 12 open reading frame 5, EC 3.1.3.46, probable fructose-2,6-bisphosphatase TIGAR, TIGARTP53-induced glycolysis and apoptosis regulator, transactivated by NS3TP2 protein
A genomic peptide made to an internal region of the human TIGAR protein (within residues 50-200). [Swiss-Prot Q9NQ88]
0.1 mL
Adaptive Immunity, Asthma, Biologically Active Proteins, Immunology, Lipid and Metabolism
57103
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.