missing translation for 'onlineSavingsMsg'
Learn More

TID1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16847933 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16847933 20 μL 20µL
16837933 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16847933

Brand: Proteintech 110881AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DNAJA3 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins. For background information on the DNAJ family.

Specifications

Antigen TID1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene DNAJA3
Gene Accession No. Q96EY1, Q99M87
Gene Alias DnaJ protein Tid 1, DNAJA3, HCA57, hTid 1, TID 1, TID1
Gene Symbols Dnaja3
Host Species Rabbit
Immunogen TID1 Fusion Protein Ag1558
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 360481, 83945, 9093
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.