Learn More
Melford Ticarcillin Disodium Salt
Description
Ticarcillin disodium salt is an antibiotic that belongs to the class of drugs known as penicillin.
Ticarcillin disodium salt is commonly used in the field of life sciences for research applications.
In the life science marketplace, ticarcillin disodium salt is primarily utilized as a selective agent in microbiology and molecular biology experiments.
It is often incorporated into culture media to inhibit the growth of certain bacteria, allowing for the selective growth of desired organisms or the detection of specific bacterial strains.
Researchers and scientists working in fields such as microbiology, immunology, and molecular biology may employ ticarcillin disodium salt to study bacterial resistance mechanisms, evaluate the efficacy of antimicrobial compounds, or isolate and identify specific bacterial species.
Specifications
Specifications
| Chemical Name or Material | Ticarcillin Disodium Salt |
| CAS | 4697-14-7 |
| Quantity | 1 g |
| Molecular Formula | C15H14N2Na2O6S2 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.