Learn More
Thymine, >99%, MP Biomedicals™
Product Code. 11453461 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453461 100 g 100g
11433461 10 g 10g
11443461 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11453461 Supplier MP Biomedicals Supplier No. 0210306080
  • ONLINE EXCLUSIVE PRICE
    4305.00 NOK
    4099.48 NOK / 100g
    Save 205.52 NOK
    4% Off
 Restricted Item
Estimated Shipment: 17-11-2026
Chemical Identifiers
CAS 65-71-4
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.115
InChI Key RWQNBRDOKXIBIV-UHFFFAOYSA-N
Synonym thymine, 5-methyluracil, thymin, 2,4-dihydroxy-5-methylpyrimidine, thymine anhydrate, 2,4 1h,3h-pyrimidinedione, 5-methyl, 5-methylpyrimidine-2,4 1h,3h-dione, 5-methyl-2,4 1h,3h-pyrimidinedione, thymin purine base, 5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
PubChem CID 1135
ChEBI CHEBI:17821
IUPAC Name 5-methyl-1H-pyrimidine-2,4-dione
SMILES CC1=CNC(=O)NC1=O
Specifications
Quantity 100 g
Formula Weight 126.1
Percent Purity >99%
Physical Form Crystalline
Chemical Name or Material Thymine