missing translation for 'onlineSavingsMsg'
Learn More

THUMPD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14921-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16883834

  • 1611.84 NOK / 20µL
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Functions as a tRNA-binding adapter to mediate NAT10-dependent tRNA acetylation.
Specifications

Specifications

THUMPD1
Polyclonal
Unconjugated
THUMPD1
THUMP domain containing 1, THUMPD1
Rabbit
Antigen Affinity Chromatography
RUO
233802, 309041, 55623
-20°C
Liquid
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q99J36, Q9NXG2
Thumpd1
THUMPD1 Fusion Protein Ag6739
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers