Learn More
Three-prong Clamps
Product Code. 12378149
Change view
Click to view available options
Diameter (Metric):
1 to 75 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
12378149 1 to 75 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12378149 Supplier S Murray SAMCO Supplier No. L001/01
  • ONLINE EXCLUSIVE PRICE
    249.00 NOK
    207.95 NOK / Pack of 1
    Save 41.05 NOK
    16% Off
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
Use this clamp for a variety of laboratory applications. Three-prong Clamps are suitable for articles from 1 to 70mm in diameter.
  • Diecast aluminum construction
  • Nickel-plated brass tommy screw
  • Rubber covered jaws
Specifications
Type Clamp
Diameter (Metric) 1 to 75 mm