Learn More
Lenz Laborglasinstrumente™ Three-Neck Flasks
Especially made for reactions and can be used to make solutions or hold, contain and collect chemicals. Lenz Laborglasinstrumente™ Three-Neck Flasks are manufactured from durable DURAN™ glass.
Brand: Lenz Laborglasinstrumente™ B0424851
Additional Details : Weight : 0.18000kg
Description
- Acc. to DIN 12392
- With two parallel necks at 90° angle
- With inscription label
- Corrosion-free and resistant to thermal shock
Specifications
250 mL | |
Three Neck | |
1 Each | |
NS 29 |