Learn More
Masterflex™ Threaded Reducing Nipple
Click to view available options
Material:
Stainless Steel (316)
Unit Size:
Each
Description
- Very good chemical resistance.
- Rated to 7500psi at 77°F (25°C).
- Usable from -65 to +550°F (-53 to +289°C).
- Threaded fittings are sturdy and designed for permanent installation.
- We offer threaded couplings and reducers, elbows and nipples, reducers and bushings, bushings, tees and plugs, and bulkhead and manifolds.
- Available in a variety of sizes and materials.
Specifications
Specifications
| Description | NPT(M) Connection |
| Material | Stainless Steel (316) |
| Sterilization Method | Autoclave, Ethylene Oxide or Gamma |
| Width (Metric) | 27 mm |
| Product Type | Threaded Reducing Nipple, 3/4 in. NPT(M) to 1/2 in. NPT(M), 316 SS |