Learn More
Masterflex™ Threaded Male Adapter Elbow
Description
- Adapts from a NPT male thread to a hose barb.
- Combine these simple and economical fittings with our flexible tubing for use in low-pressure applications (generally less than 30psi).
- Barb fittings are available in a variety of sizes, materials, and configurations from straight connectors to threaded adapter elbows to branched tees.
- A free certificate of resin compliance is available for many formulations.
- Available in a variety of materials and configurations.
Specifications
Specifications
| Description | Barb x NPT(M) |
| Material | Nylon |
| Packaging Quantity | 10/Pk. |
| Sterilization Method | Ethylene Oxide |
| Product Type | Threaded Adapter |