Learn More
Masterflex™ Thread to Compression Adapters
Click to view available options
Port Size:
NPT(M) 1/2 in.
NPT(M) 3/4 in.
Unit Size:
Pack of 10
Description
- Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal
- Available in packs of ten
Specifications
Specifications
| Description | Compression x NPT(M) Connection |
| Autoclavable | Not Autoclavable |
| Material | Polypropylene |
| Overall Length (English) | 1 15/16 in. |
| Packaging Quantity | 10 Fittings |
| Tubing Size | 3/4 in. |
| Outlet Port | 3/4 in. |
| Port Size | NPT(M) 3/4 in. |
| Sterilization Method | Ethylene Oxide |
| Width (Metric) | 33 mm |
| Show More |