Learn More
Corning™ Thread Adapters
Product Code. 16852120 Shop All Corning Products
Change view
Click to view available options
Product Type:
A32/A25
A32/A28
A32/A38
A32/A40
A32/A45
A45/A32
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
16852120 A45/A32 Each
16802120 A32/A25 Each
16812120 A32/A28 Each
16822120 A32/A38 Each
16832120 A32/A40 Each
16842120 A32/A45 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 16852120 Supplier Corning™ Supplier No. 6876
  • ONLINE EXCLUSIVE PRICE
    219.00 NOK
    203.94 NOK / Pack of 1
    Save 15.06 NOK
    6% Off
Estimated Shipment: 14-04-2027
Corning™ Thread Adapters for use with Bottle Top Dispensers
  • Threaded
  • Accessory for Bottle Top Dispenser
TRUSTED_SUSTAINABILITY
Specifications
Product Type A45/A32
For Use With (Application) Quick and precise dispensing of acids, bases and solvents
For Use With (Equipment) Bottle Top Dispenser