Learn More
SCAT™ PTFE Thread Adapters
Product Code. 10120830 Shop All SCAT Europe Products
Change view
Click to view available options
Closure Size:
GL/S40 (f) to GL45 (m)
GL/S40 (f) to S55 (m)
GL32 (f) to GL45 (m)
GL38 (f) to GL45 (m)
GL45 (f) to S55 (m)
GL45 (m) to 2 in. Tri-sure (m)
GL45 (m) to NS29/32 mm (m)
GL45 (m) to R2 in. BSP/G2 in. (m)
GPI 38-23 (f) to GL45 (m)
ND9
S51 (f) to S55 (m)
S60/61 (m) to B83 (f)
UNF 1/4 in. 28 G (f) to GL14 (m)
Unit Size:
Each
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Size Unit Size
10120830 GL38 (f) to GL45 (m) Each
10374900 GL32 (f) to GL45 (m) Each
10318729 GL45 (m) to 2 in. Tri-sure (m) Each
10829840 GL45 (m) to R2 in. BSP/G2 in. (m) Each
10840411 GL45 (f) to S55 (m) Each
10860391 GL45 (m) to NS29/32 mm (m) Each
10384420 GL/S40 (f) to GL45 (m) Each
10705802 GL/S40 (f) to S55 (m) Each
10881051 S51 (f) to S55 (m) Each
10584448 UNF 1/4 in. 28 G (f) to GL14 (m) Each
10130200 GPI 38-23 (f) to GL45 (m) Each
10671211 ND9 Each
17917270 S60/61 (m) to B83 (f) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 10120830 Leverandør SCAT™ Leverandørnr. 107992
  • ONLINE EXCLUSIVE PRICE
    842.00 NOK
    776.96 NOK / Pack of 1
    Save 65.04 NOK
    7% Off
Estimated Shipment: 28-12-2026
Læg i indkøbskurv
Læg i indkøbskurv
Denne vare kan ikke returneres. Se returpolicy
SCAT™ Safety Caps and SCAT™ Safety Waste Caps are available in many thread sizes. If the right thread for your container is still not available, simply use the matching adapter.
SCAT™ thread adapters are ideally suited for connecting safety caps, safety waste caps, funnels, canisters or other disposal and supply units.
  • Made of polytetrafluoroethylene (PTFE) material
  • Available in a wide range of thread sizes
  • Suitable for all SCAT™ caps, funnels or canisters
Tekniske data
Closure Size GL38 (f) to GL45 (m)
For Use With (Equipment) Capillary Connector
Material PTFE
Product Type Thread Adapter
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.