Learn More
Buerkle™ Thread Adapter for Mini Solvent Pump
Product Code. 15382255 Shop All Bürkle Products
Change view
Click to view available options
Tubing Size:
GL45-GL32
GL45-GL38
GL45-S40
Material:
ETFE
PTFE
Unit Size:
Each
Product Code. Tubing Size Material unitSize
15382255 GL45-GL38 ETFE Each
15302265 GL45-GL32 ETFE Each
15362255 GL45-S40 PTFE Each
3 options
Product Code. 15382255
Brand: Buerkle™ 00064002
Only null left
Add to basket
Add to basket
Thread adapter for Buerkle™ Mini solvent pump.
  • For use with Buerkle Mini Solvent Pump
  • ETFE Construction
  • External thread: GL 45
TRUSTED_SUSTAINABILITY
Specifications
Color Transparent
For Use With Mini Solvent Pump
Material ETFE
Tubing Type GL45-GL38
Tubing Size GL45-GL38
Type Thread Adapter
Weight (Metric) 0.021 kg