Learn More
SymLine™ Thread Adapter, Type 98
Shop All SCAT Europe Products
Click to view available options
Closure Size:
S60/61, S70/71
Unit Size:
Each
Description
SymLine™ offers a suitable adapter for every common thread, so that you will have no problems connecting different thread types.
- Many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables allow you to configure your SymLine™ system to your needs
- All accessories are electrostatically dissipative and approved for use in laboratories with explosion hazards
Specifications
Specifications
| Closure Size | S60/61, S70/71 |
| Closure Color | Black |
| Description | S60/61 (m) to S70/71 (f) |
| Color | Black |
| Liner Material | HDPE-EC |
| Material | HDPE-EC |
| Product Type | Thread Adapter |