Learn More
SymLine™ Angled Adapter (Type 2)
Click to view available options
Closure Size:
GL25
Unit Size:
Each
Description
SymLine™ offers a suitable adapter for every common thread, so that you will have no problems connecting different thread types.
- Many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables allow you to configure your SymLine™ system to your needs
- All accessories are electrostatically dissipative and approved for use in laboratories with explosion hazards
Specifications
Specifications
| Closure Size | GL25 |
| Closure Color | Black |
| Description | Type 2, GL25 (f) to GL25 (m) |
| Color | Black |
| Liner Material | HDPE-EC |
| For Use With (Equipment) | Desktop Mount with GL25 Connection |
| Material | HDPE-EC |
| Product Type | Adapter |