Learn More
SymLine™ Offset Adapter
Product Code. 10241850
Change view
Click to view available options
Closure Size:
GL14
Unit Size:
Each
Product Code. Closure Size unitSize
10241850 GL14 Each
1 options
Product Code. 10241850
Brand: SymLine™ 107621
Dovetail your SymLine™ System to suit your exact needs. Here, you will find many useful ancillaries.
SymLine™ offers a suitable adapter for every common thread, so that you will have no problems connecting different thread types.
  • Many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables allow you to configure your SymLine™ system to your needs
  • All accessories are electrostatically dissipative and approved for use in laboratories with explosion hazards
Specifications
Closure Size GL14
Closure Color Black
Description Extension for Exhaust Filter
Color Black
Liner Material HDPE
For Use With (Application) Attachment of Exhaust Air Filter to Waste Container in Any Position
Material HDPE
Product Type Adapter