missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THOP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11738-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Various cellular organelles and vesicles are transported along the microtubules in the cytoplasm. Likewise, membrane recycling of the endoplasmic reticulum (ER), Golgi assembly at the microtubule organizing center, and alignment of lysosomes along microtubules are all related processes. The transport of organelles requires a special class of microtubule-associated proteins (MAPs). One of these is the molecular motor kinesin (see MIM 148760 and MIM 600025), an ATPase that moves vesicles unidirectionally toward the plus end of the microtubule. Another such MAP is kinectin, a large integral ER membrane protein. Antibodies directed against kinectin have been shown to inhibit its binding to kinesin.Specifications
THOP1 | |
Polyclonal | |
Unconjugated | |
THOP1 | |
Endopeptidase 24.15, EP24.15, MEPD_HUMAN, MP78, Thimet oligopeptidase, thimet oligopeptidase 1, THOP1, TOP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
50492, 64517, 7064 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P24155, P52888, Q8C1A5 | |
Thop1 | |
THOP1 Fusion Protein Ag2268 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |