Learn More
THOC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16807633 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16807633 20 μL 20µL
16897623 150 μL 150µL
2 options
Product Code. 16807633
Brand: Proteintech 109201AP20UL
Rabbit Polyclonal Antibody
This antibody recognizes the 84 kDa THOC1 protein.
TCEB1 is known as protein elongin C, which is a subunit of the transcription factor B (SIII) complex. The SIII complex is composed of elongins A/A2, B and C. It activates elongation by RNA polymerase II by suppressing transient pausing of the polymerase at many sites within transcription units. Elongin A functions as the transcriptionally active component of the SIII complex, whereas elongins B and C are regulatory subunits. Elongin A2 is specifically expressed in the testis, and capable of forming a stable complex with elongins B and C. The von Hippel-Lindau tumor suppressor protein binds to elongins B and C, and thereby inhibits transcription elongation. Western blots using two different antibodies (P100962-T200 and P100963_P050 / P100963_P100) against two unique regions of this protein target confirm the same apparent molecular weight in our tests.
Specifications
Antigen THOC1
Applications Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene THOC1
Gene Accession No. P59924, Q8R3N6, Q96FV9
Gene Alias HPR1, hTREX84, Nuclear matrix protein p84, P84, P84N5, THO complex 1, THO complex subunit 1, Tho1, THOC1
Gene Symbols Thoc1
Host Species Rabbit
Immunogen THOC1 Fusion Protein Ag1352
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225160, 291797, 9984
Target Species Mouse, Rat, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG