Learn More
Thiosalicylic Acid, Sigma-Aldrich™
Specifications
Specifications
| CAS | 147-93-3 |
| Melting Point | 165°C to 166°C (Sublimed) |
| Density | 1.49 g/cm3 at 20°C |
| Quantity | 100 g |
| MDL Number | MFCD00004836 |
| Synonym | Thiosalicylic acid, 2-Mercaptobenzoic acid, Thiosalicylic acid |
| Molecular Weight (g/mol) | 154.19 |
| Chemical Name or Material | Thiosalicylic Acid |