missing translation for 'onlineSavingsMsg'
Learn More

Thioredoxin Rabbit anti-Human, Mouse, Rat, CoraLite 488, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech CL488-14999-100UL

 View more versions of this product

Product Code. 17639846

  • 5307.92 NOK / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Thioredoxin Polyclonal antibody apecifically detetects Thioredoxin antigen in Human, Mouse, Rat Samples. It is validated for Flow Cytometry.
Specifications

Specifications

Thioredoxin
Polyclonal
CoraLite 488
BC003377
Rabbit
Antigen affinity purification
Primary
Human, Mouse, Rat
Liquid
Flow Cytometry
1000 μg/mL
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
TXN
Thioredoxin fusion protein Ag6989
100 μL
7295
Store at -20°C. Avoid exposure to light.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.