missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thioredoxin-like 5/TRP14/TXNDC17 Antibody (043), Biotin , Novus Biologicals™
Shop All Bio Techne ProductsDescription
Thioredoxin-like 5/TRP14/TXNDC17 Monoclonal antibody specifically detects Thioredoxin-like 5/TRP14/TXNDC17 in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | Thioredoxin-like 5/TRP14/TXNDC17 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 043 |
| Conjugate | Biotin |
| Formulation | PBS |
| Gene Alias | MGC14353, Protein 42-9-9, thioredoxin (Trx)-related protein, 14 kDa, thioredoxin domain containing 17, thioredoxin domain-containing protein 17, thioredoxin-like 5, Thioredoxin-like protein 5, TRP1414 kDa thioredoxin-related protein, TXNL5 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human Thioredoxin-like 5/TRP14/TXNDC17 (Q9BRA2; Met1-Asp123). |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?