missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scharlab Thioglycolate Fluid Medium
Description
- AOAC/BAM/EP/FDA/HP/USP
- Fluid medium used for sterility testing
- According to the Eur. Pharm., USP, FDA, and for the cultivation of microaerophilic and anaerobic organisms.
Specifications
Specifications
| Certifications/Compliance | Eur. Pharm., USP, FDA |
| Form | Liquid |
| Media Type | Thioglycolate Fluid Medium |
| Product Type | Microbiology Media |
| Formulation | Peptone from Casein 15.00 g/L, Yeast Extract 5.00 g/L, Dextrose 5.50 g/L, Sodium Chloride 2.50 g/L, Sodium Thioglycolate 0.50 g/L, L-Cystine 0.50 g/L, Resazurin 0.001 g/L, Agar 0.75 g/L |
| For Use With (Application) | Sterility Testing |
| Incubation Conditions | 30°C to 35°C/20°C to 25°C (Fungi) |
| Incubation Time | ≥72 hr./ ≥5 Days (Fungi) |
| pH | 7.1 ±0.2 |
| Quantity | 500 g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?