Learn More
Thioflavine T (C.I.No.49005), Sigma-Aldrich™
Description
for synthesis
Specifications
Specifications
| CAS | 2390-54-7 |
| MDL Number | MFCD00011944 |
| Quantity | 5 g |
| Molecular Weight (g/mol) | 318.86 |
| Grade | Research |
| Chemical Name or Material | Thioflavine T (C.I.No.49005) |