Learn More
Epredia™ Thinline Biopsy Cassettes in Tube Packs
Shop All Epredia Products
Click to view available options
Color:
Beige
Blue
Gray
Green
Light Green
Lilac
Orange
Pink
Tan
White
Unit Size:
Pack of 750
Description
Printing
- Surface finish and reinforced print faces are engineered to optimize print quality
- Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
- Cassette design and flow characteristics tested to ensure efficient reagent exchange
- Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
- Precision engineering for solid connection to microtomes
Archiving
- Cassette surface texture optimizes scannability for archiving
Specifications
Specifications
| Pore Style | Small Squares |
| Closure Included | Yes |
| Quantity | 750/Cs. |
| Closure Type | Attached Lid, Front Hinge |
| PrintMate Compatible | Yes |
| Color | Gray |
| For Use With (Equipment) | For Use With PrintMate™ Printers |
| Product Type | Thinline Biopsy Cassette |