Learn More
Thiazol-4-ylboronic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Thiazol-4-ylboronic Acid |
| CAS | 1016642-07-1 |
| Molecular Formula | C3 H4 B N O2 S |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C3H4BNO2S/c6-4(7)3-1-8-2-5-3/h1-2,6-7H |
| SMILES | OB(O)c1cscn1 |
| IUPAC Name | 1,3-thiazol-4-ylboronic acid |
| Molecular Weight (g/mol) | 128.945 |
| Formula Weight | 129.006 |