Learn More
Corning™ Thermowell™ Gold 96-Well Polypropylene PCR Microplates
GREENER_CHOICE
Product Code. 10468102 Shop All Corning Products
Click to view available options
Color:
Clear
Skirt Style:
Elevated
Full
Half
Unit Size:
Pack of 50
Product Code. 10468102
Brand: Corning™ 3754
Available in full, half, and elevated skirt styles
Corning Thermowell GOLD 96-well PCR microplates are offered in four formats to ensure maximum flexibility and a perfect match for your applications. The original Thermowell 96-well PCR microplates are universal fit and can be cut into 3 x 8 well segments.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance DNase, RNase-free
Format Standard
Lid Without Lid
Array 8 x 12
Packaging 5 inner bags of 10
Color Clear
Skirt Style Elevated
Material Polypropylene
Volume (Metric) Well 200 μL
No. of Wells 96
Sterility Non-sterile
Well Shape Round
For Use With (Application) For automation, RT-PCR applications
No. per Case 50/Cs.
No. per Pack 10/Pk.
Quantity 50/Cs.
Well Color Clear