Learn More
Sonoco™ ThermoSafe PolarPack™ Standard
Product Code. 13029342
Click to view available options
Dimensions (LxWxH):
10.5 x 5.5 x 1.25 in.
10.5 x 7.5 x 1.25 in.
6 x 5.5 x 1 in.
6.5 x 5.5 x 1 in.
8 x 5.5 x 1.25 in.
Weight (English):
12 oz.
16 oz.
20 oz.
24 oz.
32 oz.
48 oz.
Material:
Polyethylene
Polyethylene Film
Unit Size:
Pack of 12
Pack of 18
Pack of 24
Pack of 36
Pack of 48
Pack of 768
Product Code. 13029342
Brand: Sonoco™ PP48
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
PolarPack refrigerants provide thermal solutions for temperature critical packages during transit. Available in a broad selection of styles and a multitude of sizes, our refrigerants are designed to provide reliable temperature protection for a vast array of temperature-sensitive products and packages.
  • Thermal solutions for temperature critical packages during transit
  • Gel will phase change at 0° C.
  • condition coolant in freezer before shipments.
  • refrigeration for transport of food, pharmaceuticals and medical products. 
  • For frozen or refrigerated shipments.
  • Alternative to wet ice.
  • Available in several sizes
  • Cold shipping supplies .
TRUSTED_SUSTAINABILITY
Specifications
Quantity 48 oz.
Length (English) Exterior 10.5 in.
Width (English) Exterior 7.5 in.
Height (English) Exterior 1.25 in.
Weight (English) 48 oz.
Dimensions (LxWxH) 10.5 x 7.5 x 1.25 in.
Material Polyethylene Film
Freezing Point 0°C